Anti-FGFR1A Mouse Monoclonal Antibody [clone: M1C4]

Supplier: Genetex
GTX20830
89288-064EA 622.08 USD
89288-064
Anti-FGFR1A Mouse Monoclonal Antibody [clone: M1C4]
Antibodies
Western blotting: at 1 ug/ml recognizes 10 ng recombinant bacterial, baculo-viral, or native FGFr1.Immunoprecipitation: Ig loop I fragment of denatured, intact FGFr1a.Immunohistochemistry: untested; predicted to be poor due to masked epitope.

Type: Primary
Antigen: FGFR1
Clonality: Monoclonal
Clone: M1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR