Anti-ANTXR1 Goat Polyclonal Antibody

Supplier: Genetex
GTX15724
89287-948EA 620.03 USD
89287-948
Anti-ANTXR1 Goat Polyclonal Antibody
Antibodies
Approx 65-70kDa band observed in Human PBMC lysates (predicted MW of 65kDa according to NP_115584). Recommended for use at 0.2-1µg/ml. This antibody is expected to recognise variant 1 (NP_115584) only.

Type: Primary
Antigen: ANTXR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Bovine, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR