Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody

Supplier: Genetex
GTX40704
89287-814EA 593.34 USD
89287-814
Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR