Anti-CK5 Mouse Monoclonal Antibody [clone: 10]

Supplier: Genetex
GTX83135
89287-476EA 630.81 USD
89287-476
Anti-CK5 Mouse Monoclonal Antibody [clone: 10]
Antibodies
Mouse monoclonal antibody [1E1] to Cytokeratin 5

Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: CK5
Clonality: Monoclonal
Clone: 10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR