Anti-CK19 Mouse Monoclonal Antibody [clone: 9H8G6]

Supplier: Genetex
GTX83113
89287-448EA 630.81 USD
89287-448
Anti-CK19 Mouse Monoclonal Antibody [clone: 9H8G6]
Antibodies
Mouse monoclonal antibody [9H8G6] to Cytokeratin 19

Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.IHC: 1/200 - 1/1000.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: CK19
Clonality: Monoclonal
Clone: 9H8G6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR