Anti-LAMB1 Mouse Monoclonal Antibody [clone: 2D9G5]

Supplier: Genetex
GTX83109
89287-442EA 630.86 USD
89287-442
Anti-LAMB1 Mouse Monoclonal Antibody [clone: 2D9G5]
Antibodies
Mouse monoclonal antibody [2D9G5] to LAMB1

Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.IHC: 1/200 - 1/1000.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: LAMB1
Clonality: Monoclonal
Clone: 2D9G5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR