Anti-TCP1B Rat Monoclonal Antibody [clone: 4E+217]

Supplier: Genetex
GTX14137
89286-130EA 622.83 USD
89286-130
Anti-TCP1B Rat Monoclonal Antibody [clone: 4E+217]
Antibodies
WB: Use at a concentration of 1 ?g/ml. Detects a band of approximately 55 kDa (predicted molecular weight: 59 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TCP1B
Clonality: Monoclonal
Clone: 4E+217
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Hamster, Monkey, Pig, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR