Anti-CD23 Rabbit Monoclonal Antibody [clone: SP23]

Supplier: Genetex
GTX79430
89285-960EA 832.75 USD
89285-960
Anti-CD23 Rabbit Monoclonal Antibody [clone: SP23]
Antibodies
Monoclonal Rabbit Antibody to Human CD23

We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: SP23
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR