Anti-PLP1 Chicken Polyclonal Antibody

Supplier: Genetex
GTX85465
89285-804EA 619.01 USD
89285-804
Anti-PLP1 Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal antibody to Proteolipid Protein (PLP) (Myelin marker)

Recommended starting dilutions: For IHC/IF: Use at 1:1000 - 1:2000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PLP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR