Anti-VAMP1 Rabbit Monoclonal Antibody [clone: SP11]

Supplier: Genetex
GTX79435
89285-152EA 661.98 USD
89285-152
Anti-VAMP1 Rabbit Monoclonal Antibody [clone: SP11]
Antibodies
Rabbit monoclonal antibody [SP11] to Synaptophysin

We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.

Type: Primary
Antigen: VAMP1
Clonality: Monoclonal
Clone: SP11
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR