Anti-SNAP25 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX82597
89283-990EA 687.83 USD
89283-990
Anti-SNAP25 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SNAP25

Type: Primary
Antigen: SNAP25
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR