Anti-SYT Rabbit Polyclonal Antibody

Supplier: Genetex
GTX82584
89283-970EA 687.83 USD
89283-970
Anti-SYT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Synaptotagmin (phospho Thr202)

Type: Primary
Antigen: SYT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR