Anti-PGR Mouse Monoclonal Antibody [clone: 608]

Supplier: Genetex
GTX30165
89283-816EA 687.83 USD
89283-816
Anti-PGR Mouse Monoclonal Antibody [clone: 608]
Antibodies
Mouse monoclonal antibody [608] to Progesterone Receptor (phospho Ser294)

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 608
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR