Anti-Calcyon (calcyon neuron-specific vesicular Protein) Chicken Polyclonal Antibody

Supplier: Genetex
GTX85123
89283-796EA 688.59 USD
89283-796
Anti-Calcyon (calcyon neuron-specific vesicular Protein) Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal antibody to Calcyon

Type: Primary
Antigen: Calcyon (calcyon neuron-specific vesicular Protein)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR