Anti-Treponema pallidum (Syphilis) Mouse Monoclonal Antibody [clone: BDI811]

Supplier: Genetex
GTX38873
89282-604EA 621.83 USD
89282-604
Anti-Treponema pallidum (Syphilis) Mouse Monoclonal Antibody [clone: BDI811]
Antibodies
Type: Primary
Antigen: Treponema pallidum (Syphilis)
Clonality: Monoclonal
Clone: BDI811
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR