Anti-MOG Mouse Monoclonal Antibody

Supplier: Genetex
GTX37234
89282-302EA 626.54 USD
89282-302
Anti-MOG Mouse Monoclonal Antibody
Antibodies
Mouse Monoclonal antibody to MOG

Type: Primary
Antigen: MOG
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR