Anti-CHFR Goat Polyclonal Antibody

Supplier: Genetex
GTX13773
89278-838EA 614.31 USD
89278-838
Anti-CHFR Goat Polyclonal Antibody
Antibodies
Preliminary experiments gave an approx 80kDa band in human brain lysates at 0.5ug/ml, this band was successfully blocked by incubation with the immunising peptide. Please note that currently we cannot find an explanation in the literature for the band we observe given the predicted size of approx. 72kDda according to NP_060693. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported? In paraffin embedded human brain (cerebellum) shows stained Purkinje cells. Recommended concentration, 2.5µg/ml. This antibody is expected to recognise all reported isoforms.

Type: Primary
Antigen: CHFR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR