Anti-CDC7 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX78149
89278-466EA 491.15 USD
89278-466
Anti-CDC7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to CDC7-Related Protein Kinase (CDC7L1)

Type: Primary
Antigen: CDC7
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR