Anti-IL10 Rat Monoclonal Antibody [clone: JES3-9D7]

Supplier: Genetex
GTX39969
89276-108EA 1018.33 USD
89276-108
Anti-IL10 Rat Monoclonal Antibody [clone: JES3-9D7]
Antibodies
Rat monoclonal antibody [JES3-9D7] to IL10

Recommended Starting Dilutions:Western Blotting: Max Dilution 1/100Flow Cytometry: 1/10 - 1/50Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: JES3-9D7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR