Anti-FERMT2 Mouse Monoclonal Antibody [clone: 25A4]

Supplier: Genetex
GTX84505
89273-796EA 719.2 USD
89273-796
Anti-FERMT2 Mouse Monoclonal Antibody [clone: 25A4]
Antibodies
Mouse monoclonal antibody [25A4] to FERMT2

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FERMT2
Clonality: Monoclonal
Clone: 25A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR