Anti-FH Mouse Monoclonal Antibody [clone: 2A2]

Supplier: Genetex
GTX84499
89273-784EA 719.2 USD
89273-784
Anti-FH Mouse Monoclonal Antibody [clone: 2A2]
Antibodies
Mouse monoclonal antibody [2A2] to FH

Recommended Starting Dilutions:WB 1:1000 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: FH
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR