Anti-GAD1 Mouse Monoclonal Antibody [clone: 3H2]

Supplier: Genetex
GTX84470
89273-730EA 719.2 USD
89273-730
Anti-GAD1 Mouse Monoclonal Antibody [clone: 3H2]
Antibodies
Mouse Monoclonal antibody [3H2] to GAD67

Recommended Starting Dilutions:WB 1:1000 IHC 1:50Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GAD1
Clonality: Monoclonal
Clone: 3H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR