Anti-GBP2 Mouse Monoclonal Antibody [clone: 1E5]

Supplier: Genetex
GTX84445
89273-680EA 719.2 USD
89273-680
Anti-GBP2 Mouse Monoclonal Antibody [clone: 1E5]
Antibodies
Mouse monoclonal antibody [1E5] to GBP2

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GBP2
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR