Anti-GLUL Mouse Monoclonal Antibody [clone: 1F4]

Supplier: Genetex
GTX84426
89273-642EA 719.83 USD
89273-642
Anti-GLUL Mouse Monoclonal Antibody [clone: 1F4]
Antibodies
Mouse monoclonal antibody [1F4] to GLUL

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GLUL
Clonality: Monoclonal
Clone: 1F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR