Anti-GSTT2 Mouse Monoclonal Antibody [clone: 2000]

Supplier: Genetex
GTX84400
89273-592EA 719.2 USD
89273-592
Anti-GSTT2 Mouse Monoclonal Antibody [clone: 2000]
Antibodies
Mouse monoclonal antibody [2E3] to GSTT2

Recommended Starting Dilutions: WB 1:500~2000 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GSTT2
Clonality: Monoclonal
Clone: 2E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR