Anti-HSD17B10 Mouse Monoclonal Antibody [clone: 11A2]

Supplier: Genetex
GTX84347
89273-486EA 719.2 USD
89273-486
Anti-HSD17B10 Mouse Monoclonal Antibody [clone: 11A2]
Antibodies
Mouse monoclonal antibody [11A2] to HSD17B10

Recommended Starting Dilutions:WB 1:1000 IHC 1:50 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: HSD17B10
Clonality: Monoclonal
Clone: 11A2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR