Anti-KLHL2 Mouse Monoclonal Antibody [clone: 6D8]

Supplier: Genetex
GTX84261
89273-316EA 719.2 USD
89273-316
Anti-KLHL2 Mouse Monoclonal Antibody [clone: 6D8]
Antibodies
Mouse monoclonal antibody [6D8] to KLHL2

Recommended Starting Dilutions: WB 1:500~2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: KLHL2
Clonality: Monoclonal
Clone: 6D8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR