Anti-MAD2L1 Mouse Monoclonal Antibody [clone: 4D2]

Supplier: Genetex
GTX84169
89273-132EA 719.2 USD
89273-132
Anti-MAD2L1 Mouse Monoclonal Antibody [clone: 4D2]
Antibodies
Mouse monoclonal antibody [4D2] to MAD2L1

Recommended Starting Dilutions:WB 1:1000 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAD2L1
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR