Anti-MAPK1 Mouse Monoclonal Antibody [clone: 4D10]

Supplier: Genetex
GTX84153
89273-100EA 719.2 USD
89273-100
Anti-MAPK1 Mouse Monoclonal Antibody [clone: 4D10]
Antibodies
Mouse monoclonal antibody [4D10] to ERK2

Recommended Starting Dilutions:WB 1:2000 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 4D10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR