Anti-PARP16 Mouse Monoclonal Antibody [clone: 2C8]

Supplier: Genetex
GTX83943
89272-684EA 719.83 USD
89272-684
Anti-PARP16 Mouse Monoclonal Antibody [clone: 2C8]
Antibodies
Mouse monoclonal antibody [2C8] to PARP16

Recommended Starting Dilutions:Flow 1:500Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PARP16
Clonality: Monoclonal
Clone: 2C8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR