Anti-PDE4A Mouse Monoclonal Antibody [clone: 7B11]

Supplier: Genetex
GTX83922
89272-644EA 719.2 USD
89272-644
Anti-PDE4A Mouse Monoclonal Antibody [clone: 7B11]
Antibodies
Mouse monoclonal antibody [7B11] to PDE4A

Recommended Starting Dilutions: WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PDE4A
Clonality: Monoclonal
Clone: 7B11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR