Anti-SATB1 Mouse Monoclonal Antibody [clone: 10G2]

Supplier: Genetex
GTX83684
89272-172EA 719.2 USD
89272-172
Anti-SATB1 Mouse Monoclonal Antibody [clone: 10G2]
Antibodies
Mouse monoclonal antibody [10G2] to SATB1

Recommended Starting Dilutions:WB 1:500 IHC 1:50 ICC/IF 1:100 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SATB1
Clonality: Monoclonal
Clone: 10G2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR