Anti-SATB1 Mouse Monoclonal Antibody [clone: 11E8]

Supplier: Genetex
GTX83679
89272-162EA 741.27 USD
89272-162
Anti-SATB1 Mouse Monoclonal Antibody [clone: 11E8]
Antibodies
Mouse monoclonal antibody [11E8] to SATB1

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SATB1
Clonality: Monoclonal
Clone: 11E8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR