Anti-SHC1 Mouse Monoclonal Antibody [clone: 1D3]

Supplier: Genetex
GTX83655
89272-114EA 719.45 USD
89272-114
Anti-SHC1 Mouse Monoclonal Antibody [clone: 1D3]
Antibodies
Mouse monoclonal antibody [1D3] to SHC1

Recommended Starting Dilutions:WB 1:2000 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SHC1
Clonality: Monoclonal
Clone: 1D3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR