Anti-ATG4 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX85759
89271-316EA 655.5 USD
89271-316
Anti-ATG4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ATG4B

WB (1 - 3 µg/ml), IHC-P (10 µg/ml)

Type: Primary
Antigen: ATG4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Chicken, Pufferfish Rat, Xenopus laevis, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR