Anti-PKDREJ Rabbit Polyclonal Antibody

Supplier: Genetex
GTX47680
89270-900EA 624.43 USD
89270-900
Anti-PKDREJ Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PKDREJ (Middle)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: PKDREJ
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR