Anti-FXYD7 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX47657
89270-866EA 624.43 USD
89270-866
Anti-FXYD7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FXYD7 (N-terminal)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: FXYD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR