Anti-CACNB2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX47534
89270-792EA 624.43 USD
89270-792
Anti-CACNB2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Cav beta 2

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CACNB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR