Anti-SFRS8 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX47368
89270-580EA 624.43 USD
89270-580
Anti-SFRS8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SFRS8 (N-terminal)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: SFRS8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR