Anti-U2SURP Rabbit Polyclonal Antibody

Supplier: Genetex
GTX47177
89270-300EA 624.43 USD
89270-300
Anti-U2SURP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to SR140 (Middle)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: U2SURP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR