Anti-CYP2A7 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX47118
89270-220EA 624.43 USD
89270-220
Anti-CYP2A7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CYP2A7 (N-terminal)

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CYP2A7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR