Anti-B4GALT2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX46701
89269-576EA 624.26 USD
89269-576
Anti-B4GALT2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to B4GALT2 (Middle)

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: B4GALT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR