Anti-C19ORF46 (chromosome 19 open reading frame 46) Rabbit Polyclonal Antibody

Supplier: Genetex
GTX46600
89269-396EA 640.14 USD
89269-396
Anti-C19ORF46 (chromosome 19 open reading frame 46) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to C19ORF46 (N-terminal)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: C19ORF46 (chromosome 19 open reading frame 46)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR