Anti-PGS1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX46171
89268-680EA 624.26 USD
89268-680
Anti-PGS1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PGS1 (C-terminal)

Recommended Dilutions: For Western Blot: Use at 0.25 ug/ml. For IHC-P: Use at 4-8 ug/mL.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: PGS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR