Anti-FAM47A Rabbit Polyclonal Antibody

Supplier: Genetex
GTX45512
89267-482EA 624.43 USD
89267-482
Anti-FAM47A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FAM47A (Middle)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: FAM47A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR