Anti-AP2B1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX45496
89267-450EA 624.43 USD
89267-450
Anti-AP2B1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to AP2B1 (Middle)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: AP2B1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR