Anti-FAM26A Rabbit Polyclonal Antibody

Supplier: Genetex
GTX45114
89266-712EA 624.43 USD
89266-712
Anti-FAM26A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FAM26A (C-terminal)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: CALHM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR