Anti-LOC554235 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX45087
89266-666EA 624.43 USD
89266-666
Anti-LOC554235 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to LOC554235 (Middle)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: LOC554235
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR