Anti-HMGCLL1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX45075
89266-642EA 640.14 USD
89266-642
Anti-HMGCLL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to HMGCLL1 (N-terminal)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: HMGCLL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR