Anti-Influenza B Mouse Monoclonal Antibody [clone: 1131 (2B2)]

Supplier: Genetex
GTX40247
89265-868EA 653.5 USD
89265-868
Anti-Influenza B Mouse Monoclonal Antibody [clone: 1131 (2B2)]
Antibodies
Type: Primary
Antigen: Influenza B
Clonality: Monoclonal
Clone: 1131 (2B2)
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR